Imprimer
Anti-ALX4 Mouse Monoclonal Antibody [clone: 2E1]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505240EA 749 EUR
ORIGCF505240
Anti-ALX4 Mouse Monoclonal Antibody [clone: 2E1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: ALX4
Clonality: Monoclonal
Clone: 2E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human