Imprimer

Anti-PARVA Mouse Monoclonal Antibody [clone: 3H2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505999EA 749 EUR
ORIGCF505999
Anti-PARVA Mouse Monoclonal Antibody [clone: 3H2]
Anticorps
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150

Type: Primary
Antigen: PARVA
Clonality: Monoclonal
Clone: 3H2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR