Imprimer
Anti-PARVA Mouse Monoclonal Antibody [clone: 2C3]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF506002EA 749 EUR
ORIGCF506002
Anti-PARVA Mouse Monoclonal Antibody [clone: 2C3]
Anticorps
Recommended Dilutions: Western Blot: 1:200-4000
Type: Primary
Antigen: PARVA
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat