Imprimer

Anti-PIK3C2B Mouse Monoclonal Antibody [clone: 2G3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF506011EA 770 EUR
ORIGCF506011
Anti-PIK3C2B Mouse Monoclonal Antibody [clone: 2G3]
Anticorps
Recommended Dilutions: Western Blot: 1:2000-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PIK3C2B
Clonality: Monoclonal
Clone: 2G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR