Imprimer
Anti-RTN4IP1 Mouse Monoclonal Antibody [clone: 1A8]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF504093EA 749 EUR
ORIGCF504093
Anti-RTN4IP1 Mouse Monoclonal Antibody [clone: 1A8]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150
Type: Primary
Antigen: RTN4IP1
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human