Imprimer
Anti-CDK2 Mouse Monoclonal Antibody [clone: 4D3]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF502893EA 749 EUR
ORIGCF502893
Anti-CDK2 Mouse Monoclonal Antibody [clone: 4D3]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: CDK2
Clonality: Monoclonal
Clone: 4D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human