Imprimer

Anti-EIF4E Mouse Monoclonal Antibody [clone: 6C3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF502896EA 764 EUR
ORIGCF502896
Anti-EIF4E Mouse Monoclonal Antibody [clone: 6C3]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: EIF4E
Clonality: Monoclonal
Clone: 6C3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR