Imprimer
Anti-PIK3CG Mouse Monoclonal Antibody [clone: 4B1]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505255EA 749 EUR
ORIGCF505255
Anti-PIK3CG Mouse Monoclonal Antibody [clone: 4B1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: PIK3CG
Clonality: Monoclonal
Clone: 4B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human