Imprimer

Anti-IFI35 Mouse Monoclonal Antibody [clone: 2H5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF503361EA 770 EUR
ORIGCF503361
Anti-IFI35 Mouse Monoclonal Antibody [clone: 2H5]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: IFI35
Clonality: Monoclonal
Clone: 2H5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR