Imprimer
Anti-KCNJ3 Mouse Monoclonal Antibody [clone: 1D3]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF504139EA 749 EUR
ORIGCF504139
Anti-KCNJ3 Mouse Monoclonal Antibody [clone: 1D3]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: KCNJ3
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human