Imprimer

Anti-SH2B3 Mouse Monoclonal Antibody [clone: 2C9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF502933EA 749 EUR
ORIGCF502933
Anti-SH2B3 Mouse Monoclonal Antibody [clone: 2C9]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SH2B3
Clonality: Monoclonal
Clone: 2C9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR