Imprimer
Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 4B7]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505337EA 786 EUR
ORIGCF505337
Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 4B7]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: CCT8L2
Clonality: Monoclonal
Clone: 4B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human