Imprimer

Anti-C2orf56 Mouse Monoclonal Antibody [clone: 4A5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF502995EA 749 EUR
ORIGCF502995
Anti-C2orf56 Mouse Monoclonal Antibody [clone: 4A5]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: C2orf56
Clonality: Monoclonal
Clone: 4A5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR