Imprimer
Anti-OSGEP Mouse Monoclonal Antibody [clone: 8B3]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF503001EA 749 EUR
ORIGCF503001
Anti-OSGEP Mouse Monoclonal Antibody [clone: 8B3]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: OSGEP
Clonality: Monoclonal
Clone: 8B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human