Imprimer
Anti-LIMK1 Mouse Monoclonal Antibody [clone: 7C3]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF503007EA 749 EUR
ORIGCF503007
Anti-LIMK1 Mouse Monoclonal Antibody [clone: 7C3]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: LIMK1
Clonality: Monoclonal
Clone: 7C3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human