Imprimer

Anti-CD1C Mouse Monoclonal Antibody [clone: 5A8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505398EA 749 EUR
ORIGCF505398
Anti-CD1C Mouse Monoclonal Antibody [clone: 5A8]
Anticorps
Recommended Dilutions: Western Blot: 1:400-4000; Immunofluorescence: 1:100

Type: Primary
Antigen: CD1C
Clonality: Monoclonal
Clone: 5A8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR