Imprimer

Anti-ARMC1 Mouse Monoclonal Antibody [clone: 2E5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505406EA 770 EUR
ORIGCF505406
Anti-ARMC1 Mouse Monoclonal Antibody [clone: 2E5]
Anticorps
Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: ARMC1
Clonality: Monoclonal
Clone: 2E5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR