Imprimer
Anti-CMPK1 Mouse Monoclonal Antibody [clone: 1H3]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505367EA 749 EUR
ORIGCF505367
Anti-CMPK1 Mouse Monoclonal Antibody [clone: 1H3]
Anticorps
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: CMPK1
Clonality: Monoclonal
Clone: 1H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human