Imprimer

Anti-EGR2 Mouse Monoclonal Antibody [clone: 1B12]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505382EA 749 EUR
ORIGCF505382
Anti-EGR2 Mouse Monoclonal Antibody [clone: 1B12]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: EGR2
Clonality: Monoclonal
Clone: 1B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR