Imprimer

Anti-PGAM2 Mouse Monoclonal Antibody [clone: 4B12]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF503440EA 749 EUR
ORIGCF503440
Anti-PGAM2 Mouse Monoclonal Antibody [clone: 4B12]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PGAM2
Clonality: Monoclonal
Clone: 4B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR