Imprimer
Anti-LGALS3BP Mouse Monoclonal Antibody [clone: 3G8]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF503455EA 749 EUR
ORIGCF503455
Anti-LGALS3BP Mouse Monoclonal Antibody [clone: 3G8]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: LGALS3BP
Clonality: Monoclonal
Clone: 3G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human