Imprimer

Anti-C20orf3 Mouse Monoclonal Antibody [clone: 4F6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504220EA 749 EUR
ORIGCF504220
Anti-C20orf3 Mouse Monoclonal Antibody [clone: 4F6]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: C20orf3
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR