Imprimer

Anti-ALOX15 Mouse Monoclonal Antibody [clone: 8C5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504249EA 764 EUR
ORIGCF504249
Anti-ALOX15 Mouse Monoclonal Antibody [clone: 8C5]
Anticorps
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ALOX15
Clonality: Monoclonal
Clone: 8C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR