Imprimer
Anti-SOCS3 Mouse Monoclonal Antibody [clone: 5C2]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF503043EA 749 EUR
ORIGCF503043
Anti-SOCS3 Mouse Monoclonal Antibody [clone: 5C2]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: SOCS3
Clonality: Monoclonal
Clone: 5C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human