Imprimer

Anti-KIF25 Mouse Monoclonal Antibody [clone: 1D3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF505424EA 770 EUR
ORIGCF505424
Anti-KIF25 Mouse Monoclonal Antibody [clone: 1D3]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: KIF25
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR