Imprimer
Anti-PCDH7 Mouse Monoclonal Antibody [clone: 3D1]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF505444EA 749 EUR
ORIGCF505444
Anti-PCDH7 Mouse Monoclonal Antibody [clone: 3D1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: PCDH7
Clonality: Monoclonal
Clone: 3D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human