Imprimer

Anti-CCM2 Mouse Monoclonal Antibody [clone: 5H4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF503462EA 749 EUR
ORIGCF503462
Anti-CCM2 Mouse Monoclonal Antibody [clone: 5H4]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CCM2
Clonality: Monoclonal
Clone: 5H4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR