Imprimer

Anti-CYP2J2 Mouse Monoclonal Antibody [clone: 5C9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF503482EA 764 EUR
ORIGCF503482
Anti-CYP2J2 Mouse Monoclonal Antibody [clone: 5C9]
Anticorps
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:500

Type: Primary
Antigen: CYP2J2
Clonality: Monoclonal
Clone: 5C9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR