Imprimer
Anti-MKRN1 Mouse Monoclonal Antibody [clone: 1G6]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF503489EA 770 EUR
ORIGCF503489
Anti-MKRN1 Mouse Monoclonal Antibody [clone: 1G6]
Anticorps
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: MKRN1
Clonality: Monoclonal
Clone: 1G6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human