Imprimer
Anti-PVRL1 Mouse Monoclonal Antibody [clone: 6F8]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF503521EA 749 EUR
ORIGCF503521
Anti-PVRL1 Mouse Monoclonal Antibody [clone: 6F8]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: PVRL1
Clonality: Monoclonal
Clone: 6F8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human