Imprimer

Anti-ERCC1 Mouse Monoclonal Antibody [clone: 1A1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504288EA 749 EUR
ORIGCF504288
Anti-ERCC1 Mouse Monoclonal Antibody [clone: 1A1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC1
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR