Imprimer

Anti-FMR1 Mouse Monoclonal Antibody [clone: 1D10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF504291EA 749 EUR
ORIGCF504291
Anti-FMR1 Mouse Monoclonal Antibody [clone: 1D10]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:50

Type: Primary
Antigen: FMR1
Clonality: Monoclonal
Clone: 1D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR