Imprimer
Anti-LENG1 Mouse Monoclonal Antibody [clone: 1G5]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF503082EA 770 EUR
ORIGCF503082
Anti-LENG1 Mouse Monoclonal Antibody [clone: 1G5]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: LENG1
Clonality: Monoclonal
Clone: 1G5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human