Imprimer
Anti-SMS Mouse Monoclonal Antibody [clone: 3C9]
Fournisseur: OriGene
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ORIGCF503099EA 749 EUR
ORIGCF503099
Anti-SMS Mouse Monoclonal Antibody [clone: 3C9]
Anticorps
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: SMS
Clonality: Monoclonal
Clone: 3C9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat