Imprimer

Anti-DOT1L Mouse Monoclonal Antibody [clone: 3E9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF802740EA 764 EUR
ORIGCF802740
Anti-DOT1L Mouse Monoclonal Antibody [clone: 3E9]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DOT1L
Clonality: Monoclonal
Clone: 3E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR