Imprimer

Anti-ZNF385B Mouse Monoclonal Antibody [clone: 4F9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF802761EA 786 EUR
ORIGCF802761
Anti-ZNF385B Mouse Monoclonal Antibody [clone: 4F9]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ZNF385B
Clonality: Monoclonal
Clone: 4F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR