Imprimer

Anti-OAS2 Mouse Monoclonal Antibody [clone: 2A1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF802773EA 786 EUR
ORIGCF802773
Anti-OAS2 Mouse Monoclonal Antibody [clone: 2A1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: OAS2
Clonality: Monoclonal
Clone: 2A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR