Imprimer

Anti-CD274 Mouse Monoclonal Antibody [clone: 2B12]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF507122EA 764 EUR
ORIGCF507122
Anti-CD274 Mouse Monoclonal Antibody [clone: 2B12]
Anticorps
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR