Imprimer

Anti-HAUS7 Mouse Monoclonal Antibody [clone: 2F1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF507054EA 764 EUR
ORIGCF507054
Anti-HAUS7 Mouse Monoclonal Antibody [clone: 2F1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: HAUS7
Clonality: Monoclonal
Clone: 2F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR