Imprimer

Anti-KI67 Mouse Monoclonal Antibody [clone: 7D9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGCF800664EA 764 EUR
ORIGCF800664
Anti-KI67 Mouse Monoclonal Antibody [clone: 7D9]
Anticorps
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: KI67
Clonality: Monoclonal
Clone: 7D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR