Imprimer

Anti-BLVRB Goat Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA327772EA 510 EUR
ORIGTA327772
Anti-BLVRB Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: BLVRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR