Imprimer

Anti-PTGDS Goat Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326303EA 510 EUR
ORIGTA326303
Anti-PTGDS Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PTGDS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR