Imprimer

Anti-SERPINH1 Mouse Monoclonal Antibody [clone: 1C4-1A6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326347EA 0 EUR
ORIGTA326347
Anti-SERPINH1 Mouse Monoclonal Antibody [clone: 1C4-1A6]
Anticorps
Type: Primary
Antigen: SERPINH1
Clonality: Monoclonal
Clone: 1C4-1A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR