Imprimer

Anti-HMOX1 Mouse Monoclonal Antibody [clone: 1F12-A6]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326354EA 649 EUR
ORIGTA326354
Anti-HMOX1 Mouse Monoclonal Antibody [clone: 1F12-A6]
Anticorps
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: HMOX1
Clonality: Monoclonal
Clone: 1F12-A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Bovine, Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR