Imprimer

Anti-HSPB2 Mouse Monoclonal Antibody [clone: 8A7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326366EA 498 EUR
ORIGTA326366
Anti-HSPB2 Mouse Monoclonal Antibody [clone: 8A7]
Recommended Dilutions: Western Blot: 1:5000; ECL 1:200

Type: Primary
Antigen: HSPB2
Clonality: Monoclonal
Clone: 8A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Bovine, canine, Guinea Pig, Hamster, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR