Imprimer

Anti-HSPD1 Mouse Monoclonal Antibody [clone: LK2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326374EA 598 EUR
ORIGTA326374
Anti-HSPD1 Mouse Monoclonal Antibody [clone: LK2]
Anticorps
Recommended Dilutions: Western Blot: 0.25 ?g/ml

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: LK2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, canine, Chicken, e.coli, Guinea Pig, h. pylori, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, s. typhimurium, spinach, t. spiralis, white fly, Yeast
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR