Imprimer

Anti-SF Rabbit Polyclonal Antibody

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326986EA 524 EUR
ORIGTA326986
Anti-SF Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:200-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: SF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR