Imprimer

Anti-Rab4A Rabbit Polyclonal Antibody [clone: N/A]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326379EA 585 EUR
ORIGTA326379
Anti-Rab4A Rabbit Polyclonal Antibody [clone: N/A]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-2000

Type: Primary
Antigen: Rab4A
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat.
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR