Imprimer

Anti-VAMP1 Mouse Monoclonal Antibody [clone: SP-11]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326386EA 485 EUR
ORIGTA326386
Anti-VAMP1 Mouse Monoclonal Antibody [clone: SP-11]
Recommended Dilutions: Western Blot: 0.5-1?g/ml

Type: Primary
Antigen: VAMP1
Clonality: Monoclonal
Clone: SP-11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human.
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR