Imprimer

Anti-AHSA1 Mouse Monoclonal Antibody [clone: 4H9-D7]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA326390EA 603 EUR
ORIGTA326390
Anti-AHSA1 Mouse Monoclonal Antibody [clone: 4H9-D7]
Anticorps
Type: Primary
Antigen: AHSA1
Clonality: Monoclonal
Clone: 4H9-D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR